Assay Details
| Gene Symbol: Slc5a4a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_133184.2 | NP_573447.2 | 13 - 14 | 1738 | 94 | |
| GenBank mRNA | AF251267.1 | - | 13 - 14 | 1738 | 94 | |
| AF498304.1 | - | 2 - 3 | 309 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Slc5a4a
Genomic Map