Assay Details
| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK082152.1 | - | 6 - 7 | 1230 | 63 | |
| AK147535.1 | - | 17 - 18 | 6818 | 63 | ||
| BC040463.1 | - | 7 - 8 | 1767 | 63 | ||
More Information
Set Membership: |
|

Cic
Genomic Map