Assay Details
| Gene Symbol: Cic | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AF363690.1 | - | 3 - 4 | 489 | 74 | |
| AK147276.1 | - | 3 - 4 | 1122 | 74 | ||
| AK147535.1 | - | 4 - 5 | 3240 | 74 | ||
More Information
Set Membership: |
|

Cic
Genomic Map