Assay Details
| Gene Symbol: Slc19a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001199271.1 | NP_001186200.1 | 1 - 2 | 41 | 54 | |
| GenBank mRNA | AK002560.1 | - | 1 - 2 | 52 | 54 | |
| AK138577.1 | - | 1 - 2 | 96 | 54 | ||
| BC015263.1 | - | 1 - 2 | 53 | 4239690 | 54 | |
| BF786711.1 | - | 1 - 2 | 52 | 54 | ||
| L36539.1 | - | 1 - 2 | 48 | 54 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK002560.1, BC015263.1, L36539.1, NM_001199271.1 transcripts.
Set Membership: |
|

Slc19a1
Genomic Map