Assay Details
| Gene Symbol: Slc5a4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_023219.2 | NP_075708.2 | 6 - 7 | 666 | 96 | |
| GenBank mRNA | AF411960.2 | - | 6 - 7 | 586 | 96 | |
| AK144296.1 | - | 609 | 96 | |||
| BC109133.1 | - | 6 - 7 | 605 | 96 | ||
| BC109134.1 | - | 6 - 7 | 605 | 96 | ||
| BC127612.1 | - | 6 - 7 | 605 | 96 | ||
| BC127615.1 | - | 6 - 7 | 605 | 40061657 | 96 | |
| BC128465.1 | - | 6 - 7 | 605 | 96 | ||
More Information
Updated Mapping Information
This assay targets the same location as RTA transcript cluster(s) TC2000000442.rn
Set Membership: |
|

Slc5a4b
Genomic Map