Assay Details
| Gene Symbol: Pbld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC038517.1 | - | 2 - 3 | 371 | 86 | |
| BC052783.1 | - | 2 - 3 | 329 | 86 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pbld1| Gene Symbol: Pbld1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | BC038517.1 | - | 2 - 3 | 371 | 86 | |
| BC052783.1 | - | 2 - 3 | 329 | 86 | ||
Set Membership: |
|