Assay Details
| Gene Symbol: Rora | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_013646.2 | NP_038674.1 | 2 - 3 | 220 | 83 | |
| GenBank mRNA | U53228.1 | - | 2 - 3 | 204 | 83 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rora
Genomic Map