Assay Details
| Gene Symbol: Irak1bp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001168240.1 | NP_001161712.1 | 1 - 2 | 530 | 134 | |
| GenBank mRNA | AK014712.1 | - | 1 - 2 | 307 | 134 | |
More Information
Updated Mapping Information
Set Membership: |
|

Irak1bp1
Genomic Map