Assay Details
| Gene Symbol: Adpgk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_028121.2 | NP_082397.1 | 3 - 4 | 71 | ||
| GenBank mRNA | AK011434.1 | - | 3 - 4 | 71 | ||
| AK149942.1 | - | 3 - 4 | 71 | |||
| AK162169.1 | - | 3 - 4 | 71 | |||
| AK167634.1 | - | 3 - 4 | 71 | |||
| AK169590.1 | - | 3 - 4 | 541 | 71 | ||
| AK204806.1 | - | 2 - 3 | 71 | |||
| BC021526.1 | - | 3 - 4 | 563 | 5353637 | 71 | |
| BC094657.1 | - | 3 - 4 | 71 | |||
More Information
Updated Mapping Information
Set Membership: |
|

Adpgk
Genomic Map