Assay Details
| Gene Symbol: Adpgk | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK149942.1 | - | 6 - 7 | 970 | 72 | |
| AK167634.1 | - | 6 - 7 | 999 | 72 | ||
| BC021526.1 | - | 6 - 7 | 985 | 5353637 | 72 | |
| BC094657.1 | - | 5 - 6 | 3678 | 72 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Adpgk
Genomic Map