| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Mouse Clariom S Array |
PSR0900008078.mm.1 |
NM_173731.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008078.mm.1 |
NM_173731.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008078.mm.1 |
TC0900001022.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900008073.mm.1 |
NM_173731.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008073.mm.1 |
NM_173731.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008073.mm.1 |
TC0900001022.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900008077.mm.1 |
NM_173731.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008077.mm.1 |
NM_173731.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008077.mm.1 |
TC0900001022.mm.1 |
MTA TC |
| Affymetrix Mouse Clariom S Array |
PSR0900008076.mm.1 |
NM_173731.2.1 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008076.mm.1 |
NM_173731.2 |
Refseq |
| Affymetrix Mouse Clariom S Array |
PSR0900008076.mm.1 |
TC0900001022.mm.1 |
MTA TC |