Assay Details
| Gene Symbol: Cntn5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001033359.2 | NP_001028531.2 | 14 - 15 | 2143 | 88 | |
| NM_001170787.1 | NP_001164258.1 | 18 - 19 | 2807 | 88 | ||
| GenBank mRNA | AK138164.1 | - | 14 - 15 | 2144 | 88 | |
| AK162856.1 | - | 14 - 15 | 2143 | 88 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Cntn5
Genomic Map