Assay Details
| Gene Symbol: Trip4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001170907.1 | NP_001164378.1 | 6 - 7 | 969 | 113 | |
| NM_019797.4 | NP_062771.2 | 9 - 10 | 1785 | 113 | ||
| GenBank mRNA | AF197574.1 | - | 6 - 7 | 844 | 113 | |
| AF539614.1 | - | 9 - 10 | 1668 | 113 | ||
| AK039024.1 | - | 6 - 7 | 854 | 113 | ||
| AK146298.1 | - | 6 - 7 | 852 | 113 | ||
| BC021316.1 | - | 6 - 7 | 857 | 5027899 | 113 | |
More Information
Updated Mapping Information
Set Membership: |
|

Trip4
Genomic Map