Assay Details
| Gene Symbol: Rab4b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_029391.2 | NP_083667.1 | 6 - 7 | 668 | 62 | |
| GenBank mRNA | AB232592.1 | - | 6 - 7 | 527 | 62 | |
| AK005314.1 | - | 6 - 7 | 668 | 62 | ||
| AK140513.1 | - | 5 - 6 | 865 | 62 | ||
| AK170552.1 | - | 6 - 7 | 682 | 62 | ||
| AK219531.1 | - | 2 - 3 | 217 | 62 | ||
| BC007147.1 | - | 6 - 7 | 612 | 3600193 | 62 | |
| BC096654.1 | - | 6 - 7 | 651 | 5056766 | 62 | |
More Information
Updated Mapping Information
Set Membership: |
|

Rab4b
Genomic Map