Assay Details
| Gene Symbol: 4933433G15Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_040719.1 | 1 - 2 | 581 | 90 | ||
| GenBank mRNA | AK014947.1 | - | 1 - 2 | 583 | 90 | |
| AK017040.1 | - | 582 | 90 | |||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | mitogen-activated protein kinase 6 | Mapk6 |
2610021I23Rik, D130053K17Rik, Erk3, Mapk4, Mapk63, Prkm4, Prkm6 |
-166.0 | Anti-sense | Chr. 9 - 75386782 - 75410018 on NCBI Build GRCm38 |
| Downstream |
Leo1, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae) |
Leo1 | Gm185 | 29.7kb | Sense | Chr. 9 - 75441524 - 75466432 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

4933433G15Rik
Genomic Map