Assay Details
| Gene Symbol: Snapc5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_183316.2 | NP_899139.2 | 2 - 3 | 209 | 111 | |
| GenBank mRNA | BC028529.1 | - | 2 - 3 | 198 | 1381169 | 111 |
| CJ165571.1 | - | 2 - 3 | 210 | 111 | ||
More Information
Set Membership: |
|

Snapc5
Genomic Map