Assay Details
| Gene Symbol: Tcf3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164147.1 | NP_001157619.1 | 4 - 5 | 613 | 71 | |
| NM_001164148.1 | NP_001157620.1 | 4 - 5 | 613 | 71 | ||
| NM_001164149.1 | NP_001157621.1 | 4 - 5 | 613 | 71 | ||
| NM_001164150.1 | NP_001157622.1 | 4 - 5 | 613 | 71 | ||
| NM_001164151.1 | NP_001157623.1 | 4 - 5 | 613 | 71 | ||
| NM_001164152.1 | NP_001157624.1 | 4 - 5 | 613 | 71 | ||
| NM_001164153.1 | NP_001157625.1 | 4 - 5 | 613 | 71 | ||
| NM_011548.4 | NP_035678.3 | 4 - 5 | 613 | 71 | ||
| GenBank mRNA | AF352579.1 | - | 4 - 5 | 308 | 71 | |
| AK017617.1 | - | 4 - 5 | 581 | 71 | ||
| AK034777.1 | - | 4 - 5 | 596 | 71 | ||
| AK038738.1 | - | 4 - 5 | 600 | 71 | ||
| AK146786.1 | - | 4 - 5 | 604 | 71 | ||
| AK147735.1 | - | 3 - 4 | 70 | |||
| AK156265.1 | - | 4 - 5 | 596 | 71 | ||
| AK161009.1 | - | 4 - 5 | 598 | 71 | ||
| AK170674.1 | - | 4 - 5 | 71 | |||
| BC006860.1 | - | 4 - 5 | 567 | 2631291 | 71 | |
| BC018260.1 | - | 3 - 4 | 238 | 4187386 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Tcf3
Genomic Map