Assay Details
| Gene Symbol: St6gal1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145933.4 | NP_666045.1 | 1 - 2 | 486 | 52 | |
| GenBank mRNA | AK084124.1 | - | 1 - 2 | 321 | 52 | |
| BC092222.1 | - | 1 - 2 | 464 | 6844069 | 52 | |
| CF898349.1 | - | 1 - 2 | 486 | 52 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK084124.1, BC092222.1, NM_145933.4 transcripts.
Set Membership: |
|

St6gal1
Genomic Map