Assay Details
| Gene Symbol: Bace1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145947.1 | NP_001139419.1 | 5 - 6 | 1333 | 71 | |
| GenBank mRNA | AK082317.1 | - | 5 - 6 | 1284 | 71 | |
More Information
Updated Mapping Information
Set Membership: |
|

Bace1
Genomic Map