Assay Details
| Gene Symbol: Gm16130 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001243265.1 | NP_001230194.1 | 3 - 4 | 951 | 59 | |
| GenBank mRNA | AK054504.1 | - | 3 - 4 | 952 | 59 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream |
cytochrome P450, family 11, subfamily a, polypeptide 1 |
Cyp11a1 |
Cyp11a, Cypxia1, D9Ertd411e, P450scc, Scc, cscc |
-6.6kb | Sense | Chr. 9 - 58015017 - 58027023 on NCBI Build GRCm38 |
| Downstream | stimulated by retinoic acid gene 6 | Stra6 | AI891933 | 29.6kb | Sense | Chr. 9 - 58063793 - 58154187 on NCBI Build GRCm38 |
More Information
Updated Mapping Information
Set Membership: |
|

Gm16130
Genomic Map