Assay Details
| Gene Symbol: Epor | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_010149.3 | NP_034279.3 | 1 - 2 | 263 | 76 | |
| GenBank mRNA | AK139031.1 | - | 1 - 2 | 162 | 76 | |
| BB842890.1 | - | 1 - 2 | 263 | 76 | ||
| BC003953.1 | - | 1 - 2 | 116 | 76 | ||
| BC046282.1 | - | 1 - 2 | 193 | 6438591 | 76 | |
| J04843.1 | - | 1 - 2 | 145 | 76 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Epor
Genomic Map