Assay Details
| Gene Symbol: Hcn4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001081192.1 | NP_001074661.1 | 5 - 6 | 1735 | 65 | |
| GenBank mRNA | BC158106.1 | - | 5 - 6 | 1835 | 9088182 | 65 |
| BC158108.1 | - | 5 - 6 | 1835 | 9088183 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Hcn4
Genomic Map