Assay Details
| Gene Symbol: Celf6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_175235.3 | NP_780444.2 | 6 - 7 | 789 | 117 | |
| GenBank mRNA | AK018246.1 | - | 5 - 6 | 586 | 117 | |
| AK034216.1 | - | 5 - 6 | 585 | 117 | ||
| BC052406.1 | - | 6 - 7 | 789 | 5716119 | 117 | |
| BC057083.1 | - | 5 - 6 | 571 | 5707546 | 117 | |
More Information
Updated Mapping Information
Set Membership: |
|

Celf6
Genomic Map