Assay Details
| Gene Symbol: Aplp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102455.1 | NP_001095925.1 | 2 - 3 | 365 | 106 | |
| NM_001102456.1 | NP_001095926.1 | 2 - 3 | 365 | 106 | ||
| NM_009691.2 | NP_033821.1 | 2 - 3 | 365 | 106 | ||
| GenBank mRNA | AK013376.1 | - | 2 - 3 | 362 | 106 | |
| AK149977.1 | - | 2 - 3 | 366 | 106 | ||
| AK150020.1 | - | 2 - 3 | 366 | 106 | ||
| AK160471.1 | - | 2 - 3 | 365 | 106 | ||
| AK164012.1 | - | 2 - 3 | 383 | 106 | ||
| AK190139.1 | - | 2 - 3 | 202 | 106 | ||
| AK192126.1 | - | 2 - 3 | 200 | 106 | ||
| AK192625.1 | - | 2 - 3 | 200 | 106 | ||
| AK192927.1 | - | 2 - 3 | 106 | |||
| BC051999.1 | - | 2 - 3 | 343 | 5716148 | 106 | |
| BC052396.1 | - | 2 - 3 | 337 | 5719918 | 106 | |
| U15571.1 | - | 2 - 3 | 341 | 106 | ||
| Z22592.1 | - | 2 - 3 | 365 | 106 | ||
| Z27070.1 | - | 2 - 3 | 363 | 106 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Aplp2
Genomic Map