Assay Details
| Gene Symbol: Ube2g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019803.3 | NP_062777.2 | 1 - 2 | 90 | 70 | |
| GenBank mRNA | AF296657.1 | - | 1 - 2 | 71 | 70 | |
| AK136365.1 | - | 1 - 2 | 77 | 70 | ||
| AK146554.1 | - | 1 - 2 | 95 | 70 | ||
| AK154464.1 | - | 1 - 2 | 111 | 70 | ||
| AK183367.1 | - | 1 - 2 | 43 | 70 | ||
| AK185288.1 | - | 1 - 2 | 33 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ube2g2
Genomic Map