Assay Details
| Gene Symbol: Ube2g2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019803.3 | NP_062777.2 | 2 - 3 | 134 | 93 | |
| GenBank mRNA | AF296657.1 | - | 2 - 3 | 115 | 93 | |
| AK136365.1 | - | 2 - 3 | 121 | 93 | ||
| AK146554.1 | - | 2 - 3 | 139 | 93 | ||
| AK154464.1 | - | 2 - 3 | 155 | 93 | ||
| AK183367.1 | - | 2 - 3 | 87 | 93 | ||
| BC010321.1 | - | 2 - 3 | 134 | 2651046 | 93 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ube2g2
Genomic Map