Assay Details
| Gene Symbol: Siae | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_011734.3 | NP_035864.2 | 3 - 4 | 411 | 70 | |
| GenBank mRNA | AF156856.1 | - | 4 - 5 | 613 | 70 | |
| AK028598.1 | - | 3 - 4 | 455 | 70 | ||
| AK028656.1 | - | 3 - 4 | 411 | 70 | ||
| AK035715.1 | - | 1 - 2 | 145 | 70 | ||
| AK043392.1 | - | 3 - 4 | 432 | 70 | ||
| AK158634.1 | - | 2 - 3 | 349 | 70 | ||
| AK164852.1 | - | 3 - 4 | 418 | 70 | ||
| AK167103.1 | - | 3 - 4 | 427 | 70 | ||
| BC007136.1 | - | 3 - 4 | 429 | 3592286 | 70 | |
| U40408.1 | - | 3 - 4 | 417 | 70 | ||
| U61183.1 | - | 3 - 4 | 410 | 70 | ||
| X98625.1 | - | 3 - 4 | 410 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Siae
Genomic Map