Assay Details
| Gene Symbol: Itpkc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181593.2 | NP_853624.1 | 3 - 4 | 1565 | 61 | |
| GenBank mRNA | AK033605.1 | - | 3 - 4 | 1234 | 61 | |
| AK154888.1 | - | 3 - 4 | 1597 | 61 | ||
| AK199441.1 | - | 1 - 2 | 87 | 61 | ||
| BC013227.1 | - | 3 - 4 | 1451 | 61 | ||
| BC053450.1 | - | 3 - 4 | 1565 | 6508198 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itpkc
Genomic Map