Assay Details
| Gene Symbol: Itpkc | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181593.2 | NP_853624.1 | 5 - 6 | 1874 | 91 | |
| GenBank mRNA | AK033605.1 | - | 5 - 6 | 1543 | 91 | |
| AK154888.1 | - | 5 - 6 | 1906 | 91 | ||
| BC013227.1 | - | 5 - 6 | 1760 | 91 | ||
| BC053450.1 | - | 5 - 6 | 1874 | 6508198 | 91 | |
More Information
Updated Mapping Information
Set Membership: |
|

Itpkc
Genomic Map