Assay Details
| Gene Symbol: Polr2i | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027259.1 | NP_081535.1 | 3 - 4 | 462 | 68 | |
| GenBank mRNA | AB221537.1 | - | 3 - 4 | 190 | 68 | |
| AK012635.1 | - | 3 - 4 | 199 | 68 | ||
| BC062812.1 | - | 3 - 4 | 462 | 3466417 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Polr2i
Genomic Map