Assay Details
| Gene Symbol: 9530077C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026739.1 | NP_081015.1 | 4 - 5 | 1144 | 66 | |
| GenBank mRNA | AK210022.1 | - | 1 - 2 | 147 | 66 | |
| AK220359.1 | - | 4 - 5 | 1208 | 66 | ||
| BC039998.1 | - | 2 - 3 | 367 | 3326047 | 66 | |
| BC054761.1 | - | 4 - 5 | 1144 | 6413638 | 66 | |
More Information
Updated Mapping Information
Set Membership: |
|

9530077C05Rik
Genomic Map