Assay Details
| Gene Symbol: 9530077C05Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_026739.1 | NP_081015.1 | 5 - 6 | 1451 | 63 | |
| GenBank mRNA | AK170604.1 | - | 3 - 4 | 3184 | 63 | |
| AK220359.1 | - | 5 - 6 | 1515 | 63 | ||
| BC039998.1 | - | 3 - 4 | 674 | 3326047 | 63 | |
| BC054761.1 | - | 5 - 6 | 1451 | 6413638 | 63 | |
More Information
Set Membership: |
|

9530077C05Rik
Genomic Map