Assay Details
| Gene Symbol: Pknox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029838.2 | NP_001025009.1 | 11 - 12 | 1297 | 68 | |
| NM_148950.3 | NP_683752.2 | 10 - 11 | 1192 | 68 | ||
| GenBank mRNA | AB086817.1 | - | 8 - 9 | 1018 | 68 | |
| AF487460.1 | - | 9 - 10 | 1098 | 68 | ||
| AK082952.1 | - | 11 - 12 | 1242 | 68 | ||
| BC050865.1 | - | 10 - 11 | 1126 | 6332968 | 68 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pknox2
Genomic Map