Assay Details
| Gene Symbol: Pknox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029838.2 | NP_001025009.1 | 4 - 5 | 367 | 70 | |
| NM_148950.3 | NP_683752.2 | 3 - 4 | 262 | 70 | ||
| GenBank mRNA | AB086817.1 | - | 1 - 2 | 88 | 70 | |
| AK082952.1 | - | 4 - 5 | 312 | 70 | ||
| AK084178.1 | - | 4 - 5 | 310 | 70 | ||
| AK144270.1 | - | 4 - 5 | 304 | 70 | ||
| BC050865.1 | - | 3 - 4 | 196 | 6332968 | 70 | |
| BY346714.1 | - | 3 - 4 | 207 | 70 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Pknox2
Genomic Map