Assay Details
| Gene Symbol: Pknox2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001029838.2 | NP_001025009.1 | 7 - 8 | 867 | 106 | |
| NM_148950.3 | NP_683752.2 | 6 - 7 | 762 | 106 | ||
| GenBank mRNA | AB086817.1 | - | 4 - 5 | 588 | 106 | |
| AF487460.1 | - | 5 - 6 | 668 | 106 | ||
| AK082952.1 | - | 7 - 8 | 812 | 106 | ||
| BC050865.1 | - | 6 - 7 | 696 | 6332968 | 106 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pknox2
Genomic Map