Assay Details
| Gene Symbol: Esam | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_027102.3 | NP_081378.1 | 2 - 3 | 389 | 94 | |
| GenBank mRNA | AF361882.1 | - | 2 - 3 | 374 | 94 | |
| AK009223.1 | - | 2 - 3 | 365 | 94 | ||
| AK156399.1 | - | 2 - 3 | 387 | 94 | ||
| BC019460.1 | - | 2 - 3 | 373 | 4188050 | 94 | |
| BC049249.1 | - | 2 - 3 | 369 | 5039892 | 94 | |
| BQ043033.1 | - | 2 - 3 | 330 | 94 | ||
| CJ048383.1 | - | 2 - 3 | 371 | 94 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Esam
Genomic Map