Assay Details
| Gene Symbol: Ptpn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019651.2 | NP_062625.2 | 5 - 6 | 1155 | 105 | |
| GenBank mRNA | AK089069.1 | - | 5 - 6 | 729 | 105 | |
| BC053017.1 | - | 5 - 6 | 1155 | 5708923 | 105 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ptpn9
Genomic Map