Assay Details
| Gene Symbol: Ptpn9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_019651.2 | NP_062625.2 | 6 - 7 | 1260 | 73 | |
| GenBank mRNA | BC053017.1 | - | 6 - 7 | 1260 | 5708923 | 73 |
More Information
Set Membership: |
|

Ptpn9
Genomic Map