Assay Details
| Gene Symbol: Scamp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022813.3 | NP_073724.1 | 2 - 3 | 243 | 61 | |
| GenBank mRNA | AF295402.1 | - | 2 - 3 | 247 | 61 | |
| AK003080.1 | - | 2 - 3 | 244 | 61 | ||
| AK161002.1 | - | 2 - 3 | 243 | 61 | ||
| AK171766.1 | - | 2 - 3 | 234 | 61 | ||
| BC003452.1 | - | 2 - 3 | 217 | 61 | ||
| BC014751.1 | - | 2 - 3 | 220 | 4235556 | 61 | |
More Information
Updated Mapping Information
Set Membership: |
|

Scamp2
Genomic Map