Assay Details
| Gene Symbol: Ppp2r1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034085.2 | NP_001029257.1 | 2 - 3 | 257 | 74 | |
| NM_001286553.1 | NP_001273482.1 | 2 - 3 | 257 | 74 | ||
| NM_028614.3 | NP_082890.2 | 2 - 3 | 257 | 74 | ||
| GenBank mRNA | AK077158.1 | - | 2 - 3 | 211 | 74 | |
| AK136106.1 | - | 2 - 3 | 215 | 74 | ||
| AK161392.1 | - | 2 - 3 | 210 | 74 | ||
| BC056218.1 | - | 2 - 3 | 251 | 30016074 | 74 | |
| BY722866.1 | - | 2 - 3 | 258 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r1b
Genomic Map