Assay Details
| Gene Symbol: Ppp2r1b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001034085.2 | NP_001029257.1 | 8 - 9 | 1086 | 84 | |
| NM_001286553.1 | NP_001273482.1 | 8 - 9 | 1086 | 84 | ||
| NM_028614.3 | NP_082890.2 | 8 - 9 | 1086 | 84 | ||
| GenBank mRNA | AK077158.1 | - | 8 - 9 | 1040 | 84 | |
| AK136106.1 | - | 8 - 9 | 1042 | 84 | ||
| AK161392.1 | - | 8 - 9 | 1039 | 84 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Ppp2r1b
Genomic Map