Assay Details
| Gene Symbol: Dlat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145614.4 | NP_663589.3 | 10 - 11 | 1699 | 65 | |
| GenBank mRNA | AK032124.1 | - | 10 - 11 | 1700 | 65 | |
| AK043771.1 | - | 10 - 11 | 1388 | 65 | ||
| AK166090.1 | - | 10 - 11 | 1469 | 65 | ||
| AY044265.1 | - | 10 - 11 | 1136 | 65 | ||
| BC026680.1 | - | 10 - 11 | 1447 | 5059786 | 65 | |
| BC031495.1 | - | 10 - 11 | 1420 | 2651095 | 65 | |
| BC069862.1 | - | 10 - 11 | 1442 | 5388446 | 65 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dlat
Genomic Map