Assay Details
| Gene Symbol: Dlat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145614.4 | NP_663589.3 | 11 - 12 | 1810 | 65 | |
| GenBank mRNA | AK032124.1 | - | 11 - 12 | 1811 | 65 | |
| AK043771.1 | - | 11 - 12 | 1499 | 65 | ||
| AK166090.1 | - | 11 - 12 | 1580 | 65 | ||
| AY044265.1 | - | 11 - 12 | 1247 | 65 | ||
| BC026680.1 | - | 11 - 12 | 1558 | 5059786 | 65 | |
| BC069862.1 | - | 11 - 12 | 1553 | 5388446 | 65 | |
More Information
Set Membership: |
|

Dlat
Genomic Map