Assay Details
| Gene Symbol: Dlat | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_145614.4 | NP_663589.3 | 5 - 6 | 1094 | 94 | |
| GenBank mRNA | AK032124.1 | - | 5 - 6 | 1095 | 94 | |
| AK043771.1 | - | 5 - 6 | 783 | 94 | ||
| AK166090.1 | - | 5 - 6 | 864 | 94 | ||
| AK206834.1 | - | 2 - 3 | 151 | 94 | ||
| BC026680.1 | - | 5 - 6 | 842 | 5059786 | 94 | |
| BC031495.1 | - | 5 - 6 | 815 | 2651095 | 94 | |
| BC069862.1 | - | 5 - 6 | 837 | 5388446 | 94 | |
More Information
Updated Mapping Information
Set Membership: |
|

Dlat
Genomic Map