Assay Details
| Gene Symbol: Bckdhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK089921.1 | - | 1 - 2 | 214 | 74 | |
| AK156264.1 | - | 1 - 2 | 211 | 74 | ||
| AV455466.1 | - | 1 - 2 | 224 | 74 | ||
| L16992.1 | - | 1 - 2 | 208 | 74 | ||
More Information
Set Membership: |
|

Bckdhb
Genomic Map