Assay Details
| Gene Symbol: Bckdhb | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_199195.1 | NP_954665.1 | 3 - 4 | 275 | 98 | |
| GenBank mRNA | AK089921.1 | - | 3 - 4 | 353 | 98 | |
| AK156264.1 | - | 3 - 4 | 350 | 98 | ||
| AV455466.1 | - | 3 - 4 | 363 | 98 | ||
| BC064099.1 | - | 3 - 4 | 275 | 1246285 | 98 | |
| L16992.1 | - | 3 - 4 | 347 | 98 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Bckdhb
Genomic Map