Assay Details
| Gene Symbol: Chordc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_025844.2 | NP_080120.1 | 4 - 5 | 457 | 129 | |
| GenBank mRNA | AK003259.1 | - | 4 - 5 | 458 | 129 | |
| AK012747.1 | - | 4 - 5 | 442 | 129 | ||
| AK132696.1 | - | 437 | 129 | |||
| BC018374.1 | - | 4 - 5 | 457 | 4236535 | 129 | |
More Information
Updated Mapping Information
Set Membership: |
|

Chordc1
Genomic Map