Assay Details
| Gene Symbol: Pinlyp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001037143.2 | NP_001032220.1 | 2 - 3 | 203 | 69 | |
| GenBank mRNA | AK009589.1 | - | 2 - 3 | 208 | 69 | |
| AK009604.1 | - | 2 - 3 | 205 | 69 | ||
| AK009778.1 | - | 2 - 3 | 203 | 69 | ||
| AK009924.1 | - | 2 - 3 | 205 | 69 | ||
| BC139039.1 | - | 2 - 3 | 269 | 8862061 | 69 | |
| BC145686.1 | - | 2 - 3 | 269 | 40130989 | 69 | |
More Information
Updated Mapping Information
Set Membership: |
|

Pinlyp
Genomic Map