Assay Details
| Gene Symbol: Dedd2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_207677.3 | NP_997560.3 | 4 - 5 | 737 | 74 | |
| GenBank mRNA | AF457576.1 | - | 3 - 4 | 602 | 74 | |
| AF543541.1 | - | 3 - 4 | 602 | 74 | ||
| AK010701.1 | - | 4 - 5 | 549 | 74 | ||
| AK011713.1 | - | 3 - 4 | 443 | 74 | ||
| BC092250.1 | - | 4 - 5 | 737 | 5043318 | 74 | |
| CX201206.1 | - | 3 - 4 | 589 | 74 | ||
More Information
Updated Mapping Information
Set Membership: |
|

Dedd2
Genomic Map