Assay Details
| Gene Symbol: Ubash3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_176860.5 | NP_789830.1 | 10 - 11 | 73 | ||
| GenBank mRNA | AB075602.1 | - | 10 - 11 | 73 | ||
| AK013361.1 | - | 10 - 11 | 73 | |||
| AK034450.1 | - | 10 - 11 | 73 | |||
| AK035764.1 | - | 10 - 11 | 73 | |||
| AK133948.1 | - | 10 - 11 | 73 | |||
| AK150909.1 | - | 10 - 11 | 73 | |||
| AK151721.1 | - | 10 - 11 | 73 | |||
| AK151895.1 | - | 10 - 11 | 73 | |||
| AK152013.1 | - | 10 - 11 | 73 | |||
| AK154576.1 | - | 10 - 11 | 1712 | 73 | ||
| BC053436.1 | - | 10 - 11 | 1462 | 6510702 | 73 | |
More Information
Updated Mapping Information
Set Membership: |
|

Ubash3b
Genomic Map